Learn More
actin, alpha 2, smooth muscle, aorta, Mouse, Clone: 1D11-2B9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ACTA2.
Supplier: Abnova Corporation H00000059M01
Description
The protein encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Defects in this gene cause aortic aneurysm familial thoracic type 6. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: MCEEEDSTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKQEYDEAGPSIVHRKCFSpecifications
| actin, alpha 2, smooth muscle, aorta | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC017554 | |
| ACTA2 | |
| ACTA2 (AAH17554.1, 1 a.a. ∼ 377 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 59 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 1D11-2B9 | |
| Mouse monoclonal antibody raised against a full-length recombinant ACTA2. | |
| ACTA2 | |
| AAT6/ACTSA | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |