Learn More
Acetyl-Histone H3 (Lys27) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma 07360
 View more versions of this product
Catalog No. 07360MI
Only null left
Description
Description
Anti-acetyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated on lysine 27. Also known as Anti-H3K27ac this antibody is published in peer reviewed journals and is specificity verified by dot blot (DB) and validated in Chimmunoprecipitation, ChIP-seq, western blot, DB, Mplex.
Specifications
Chemical Identifiers
acetyl-Histone H3 (Lys27)
Unconjugated
Rabbit
Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, and 5mg/ml BSA before the addition of glycerol to 30%
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
IgG
Unpurified
Polyclonal
Serum
Human, S. cerevisiae, Vertebrates, Yeast
NM_003493
synthetic peptide corresponding to the sequence ARACKSA, in which ACK corresponds to acetyl lysine 27 of yeast histone H3
Primary
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Specifications
acetyl-Histone H3 (Lys27)
Polyclonal
Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, and 5mg/ml BSA before the addition of glycerol to 30%
Rabbit
Unpurified
Primary
Human, S. cerevisiae, Vertebrates, Yeast
Serum
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Western Blot
Unconjugated
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
synthetic peptide corresponding to the sequence ARACKSA, in which ACK corresponds to acetyl lysine 27 of yeast histone H3
100 μL
NM_003493
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
IgG
Product Suggestions
Product Suggestions