missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-Histone H3 (Lys23) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

Supplier:  MilliporeSigma 07355S

 View more versions of this product

Catalog No. 07355SMI

Only null left

Description

Description

Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, western blotting to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).
Specifications

Chemical Identifiers

acetyl-Histone H3 (Lys23)
Polyclonal
Serum
Human, S. cerevisiae, Vertebrates, Yeast
NM_003493
Ovalbumin-conjugated, synthetic peptide (KQLASAcKAARK-C) corresponding to amino acids 18-27 of yeast histone H3 acetylated on lysine 23 with a C-terminal cysteine added for conjugation purposes
Antiserum
ChIP Assay, Western Blot
Unconjugated
Rabbit
Rabbit antiserum containing 0.05% sodium azide and 30% glycerol
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Primary
−20°C for 24 months

Specifications

acetyl-Histone H3 (Lys23)
Polyclonal
Rabbit antiserum containing 0.05% sodium azide and 30% glycerol
Rabbit
Antiserum
Primary
Human, S. cerevisiae, Vertebrates, Yeast
Serum
ChIP Assay, Western Blot
Unconjugated
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Ovalbumin-conjugated, synthetic peptide (KQLASAcKAARK-C) corresponding to amino acids 18-27 of yeast histone H3 acetylated on lysine 23 with a C-terminal cysteine added for conjugation purposes
10 μL
NM_003493
−20°C for 24 months
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only