Learn More
Acetyl-Histone H3 (Lys18) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma 07354S
 View more versions of this product
Catalog No. 07354SMI
Only null left
Description
Description
Use Anti-acetyl-Histone H3 (Lys18) Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, western blotting to detect acetyl-Histone H3 (Lys18) also known as H3K18Ac, Histone H3 (acetyl K18).
Specifications
Chemical Identifiers
acetyl-Histone H3 (Lys18)
Polyclonal
Serum
Human, S. cerevisiae, Vertebrates, Yeast
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Primary
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
ChIP Assay, Western Blot
Unconjugated
Rabbit
NM_003493
KLH-conjugated, synthetic peptide (GKAPRAcKQLASK-C) corresponding to amino acids 13-23 of yeast Histone H3 acetylated on lysine 18with a C-terminal cysteine added for conjugation purposes
Unpurified
Specifications
acetyl-Histone H3 (Lys18)
Polyclonal
10μLof rabbit serum containing 0.05% sodium azide and 30% glycerol.
Rabbit
Unpurified
Primary
Human, S. cerevisiae, Vertebrates, Yeast
Serum
ChIP Assay, Western Blot
Unconjugated
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
KLH-conjugated, synthetic peptide (GKAPRAcKQLASK-C) corresponding to amino acids 13-23 of yeast Histone H3 acetylated on lysine 18with a C-terminal cysteine added for conjugation purposes
10 μL
NM_003493
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Product Suggestions
Product Suggestions