missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ABP1.
Supplier: Abnova Corporation H00000026A01
Description
This gene encodes a membrane glycoprotein that is expressed in many epithelium-rich and/or hematopoietic tissues and oxidatively deaminates putrescine and histamine. The protein may play a role in controlling the level of histamine and/or putrescine in these tissues. It also binds to and is inhibited by amiloride, a diuretic that acts by closing epithelial sodium ion channels. [provided by RefSeq
Sequence: LHTHLIGNIHTHLVHYRVDLDVAGTKNSFQTLQMKLENITNPWSPRHRVVQPTLEQTQYSWERQAAFRFKRKLPKYLLFTSPQENPWGHKRSYRLQIHSSpecifications
| ABP1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ABP1. | |
| ABP1 | |
| ABP/AOC1/DAO/DAO1/KAO | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001091 | |
| ABP1 | |
| ABP1 (NP_001082, 501 a.a. ∼ 599 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 26 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |