missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA1 Mouse anti-Chicken, Human, Mouse, Clone: AB.H10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB10005
Description
This Anti-ABCA1 Antibody, clone AB.H10 is validated for use in ELISA, immunoprecipitation, western blot, immunohistochemistry (paraffin) for the detection of ABCA1.Specifications
| ABCA1 | |
| Monoclonal | |
| Unconjugated | |
| NM_005502.2 | |
| Mouse | |
| 100 μg | |
| Chicken, Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| AB.H10 | |
| Presented as a liquid in PBS, pH 7.4 with 0.05% sodium azide. | |
| ABCA1, MGC165011, HDLDT1, ABC1, TGD, CERP, membrane-bound, TD, ABC-1, MGC164864, FLJ14958 | |
| Recombinant fragment, corresponding to amino acids 1800-2260 of Human ABCA1. | |
| Primary | |
| −20°C for up to 12 months. | |
| IgG1 |
For Research Use Only