missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-6xHis Monoclonal Antibody (6A12) , TCI America™
Anti-6xHis Monoclonal Antibody (6A12), TCI America™
Supplier: TCI America A29571VIAL
Specifications
| 6xHis Monoclonal | |
| Monoclonal | |
| This monoclonal antibody was raised against a recombinant protein containing the sequence HHHHHH(6xHis) | |
| Primary | |
| Purified |
| ELISA, Western Blot | |
| Mouse | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgG1 |