missing translation for 'onlineSavingsMsg'
Learn More
Learn More
20S proteasome subunit Β6 Rabbit anti-Human, Mouse, Rabbit, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 09279
Description
Detect 20S proteasome subunit β6 using this Anti-20S proteasome subunit β6 Antibody validated for use in western blot, immunohistochemistry.Specifications
| 20S proteasome subunit β6 | |
| Polyclonal | |
| 100μL of purified antiserum in PBS containing 0.01M sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4. | |
| Rabbit | |
| Serum | |
| Primary | |
| Human, Mouse, Rabbit | |
| Serum |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| PSMB1; FLJ25321; HC5; KIAA1838; PSC5; PMSB1 | |
| Peptide corresponding to residues 185-198 of human proteasome subunit β6 conjugated to KLH. | |
| 100 μL | |
| NM_002793.2 | |
| -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles | |
| IgG |
For Research Use Only