missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 Ε Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™

Catalog No. ab9732mi
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9732MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9732MI Supplier MilliporeSigma Supplier No. AB9732
Only null left

Rabbit Polyclonal Antibody

Anti-14-3-3 Ε Antibody, C-terminal is an antibody against 14-3-3 Ε for use in ELISA, western blot, immunohistochemistry.

Specifications

Antigen 14-3-3 ε C Terminal
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid.
Gene Symbols YWHAE; KCIP-1; MDS; 14-3-3E; MDCR; FLJ45465
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminal area of 14-3-3 epsilon.
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006761.3
Target Species Bovine, Chicken, Human, Mouse, Rabbit, Rat, Sheep
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only