missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, ACS, Spectrum™ Chemical

Catalog No. p-7165114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608893 5 g Amber Glass Bottle
18608892 25 g Amber Glass Bottle
18608891 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608893 Supplier Spectrum Chemical Mfg Cor Supplier No. AN1355GM
Only null left

Chemical Identifiers

CAS 90-44-8

Specifications

CAS Max % 1
Melting Point Within 5°C
Packaging Amber Glass Bottle
Quantity 5 g
Grade ACS
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.