missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, ACS reagent

Catalog No. AC104960250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104960250 25 g Plastic bottle
AC104961000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104960250 Supplier Thermo Scientific Chemicals Supplier No. 104960250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-44-8 | C14H10O | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 10H-anthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Absorbance 0.75 max. (425nm, 1cm cell) Passes Test, 0.045 max. (620nm, 1cm cell) Passes Test
Melting Point 152.0°C to 157.0°C
Color Yellow
Boiling Point 721.0°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 25 g
Beilstein 07,473
Sensitivity (to carbohydrates) Passes Test
Merck Index 15,682
Solubility Information (in ethyl acetate) Passes test
Formula Weight 194.23
Grade ACS Reagent
Physical Form Crystals
Chemical Name or Material Anthrone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-994-

RUO – Research Use Only