missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthrone, ≥97% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 90-44-8 |
| Physical Form | Neat |
| Linear Formula | C14H10 O |
| MDL Number | MFCD00001187 |
| Melting Point | 154°C to 157°C |
| Synonym | 9(10 H)-Anthracenone |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 194.23 |
| Percent Purity | ≥97% (GC) |
| Show More |