Learn More
Anthrone, 95%
CAS: 90-44-8 | C14H10O | 194.23 g/mol
Supplier: Thermo Scientific Chemicals A1911836
Description
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.
Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Anthrone | |
| 90-44-8 | |
| C14H10O | |
| 500 g | |
| Light sensitive | |
| anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
| RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
| 9,10-dihydroanthracen-9-one | |
| 7018 | |
| 194.23 | |
| Crystalline Powder |
| 98% | |
| 153°C to 160°C | |
| MFCD00001187 | |
| 1910173 | |
| 14,691 | |
| Soluble in alcohol, benzene, and hot NaOH solutions; Insoluble in water | |
| O=C1C2=CC=CC=C2CC2=CC=CC=C12 | |
| 194.23 | |
| CHEBI:33835 | |
| 97% |
Chemical Identifiers
| 90-44-8 | |
| 194.23 | |
| RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
| 7018 | |
| O=C1C2=CC=CC=C2CC2=CC=CC=C12 |
| C14H10O | |
| MFCD00001187 | |
| anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
| CHEBI:33835 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-994-0
RTECSNumber : CB8925500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only