missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, 95%

Catalog No. AAA1911814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1911814 25 g
AAA1911822 100 g
AAA1911836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1911814 Supplier Thermo Scientific Chemicals Supplier No. A1911814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 90-44-8 | C14H10O | 194.23 g/mol

It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
PubChem CID 7018
ChEBI CHEBI:33835
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Melting Point 153°C to 160°C
Color Yellow
Quantity 25 g
Beilstein 1910173
Sensitivity Light sensitive
Merck Index 14,691
Solubility Information Soluble in alcohol, benzene, and hot NaOH solutions; Insoluble in water
IUPAC Name 10H-anthracen-9-one
Formula Weight 194.23
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Anthrone, Alfa Aesar, 98%
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-994-0
RTECSNumber CB8925500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only