missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone-2,3-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. A16811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16811G Supplier TCI America Supplier No. A16811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27485-15-0
Molecular Formula C16H8O6
Molecular Weight (g/mol) 296.23
MDL Number MFCD00087537
InChI Key PSHVQIKBLXBIQJ-UHFFFAOYSA-N
PubChem CID 3678091
IUPAC Name 9,10-dioxo-9,10-dihydroanthracene-2,3-dicarboxylic acid
SMILES OC(=O)C1=C(C=C2C(=O)C3=CC=CC=C3C(=O)C2=C1)C(O)=O

Specifications

Melting Point 342°C
Quantity 1 g
Formula Weight 296.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Anthraquinone-2,3-dicarboxylic Acid