missing translation for 'onlineSavingsMsg'
Learn More

Anthralin (Dithranol) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012643
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012643 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012643 Supplier Merck Emd Millipore Supplier No. PHR16771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Anthralin (Dithranol) is the synthetic form of chrysarobin. It is widely used as a medication for the human skin affected with psoriasis and in short contact therapy to minimize the staining, irritation, etc.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 1143-38-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C14H10O3
MDL Number MFCD00053409
Melting Point 178°C to 181°C
Synonym Dithranol; 1,8,9-Anthracenetriol; 1,8-Dihydroxy-9(10 H)-anthracenone; 1,8-Dihydroxyanthrone; Anthralin
Type Certified Reference Material
Quantity 1 g
Formula Weight 226.23
Grade Certified Reference Material