missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthraflavic Acid 97.0+%, TCI America™
Supplier: TCI America A189425G
Specifications
| Anthraflavic Acid | |
| 330°C | |
| 25 g | |
| APAJFZPFBHMFQR-UHFFFAOYSA-N | |
| 2,6-dihydroxyanthracene-9,10-dione | |
| 6776 | |
| 240.21 | |
| Crystalline Powder |
| 84-60-6 | |
| C14H8O4 | |
| 2,6-dihydroxyanthraquinone, anthraflavic acid, anthraflavin, 2,6-dihydroxyanthra-9,10-quinone, 9,10-anthracenedione, 2,6-dihydroxy, az-f, anthrafravic acid, anthraquinone, 2,6-dihydroxy, 2,6-dihydroxy-9,10-anthraquinone, 2,6-dihydroxy-anthraquinone | |
| C1=CC2=C(C=C1O)C(=O)C3=C(C2=O)C=C(C=C3)O | |
| 240.214 | |
| CHEBI:34250 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 84-60-6 | |
| 240.214 | |
| 2,6-dihydroxyanthraquinone, anthraflavic acid, anthraflavin, 2,6-dihydroxyanthra-9,10-quinone, 9,10-anthracenedione, 2,6-dihydroxy, az-f, anthrafravic acid, anthraquinone, 2,6-dihydroxy, 2,6-dihydroxy-9,10-anthraquinone, 2,6-dihydroxy-anthraquinone | |
| CHEBI:34250 | |
| C1=CC2=C(C=C1O)C(=O)C3=C(C2=O)C=C(C=C3)O |
| C14H8O4 | |
| APAJFZPFBHMFQR-UHFFFAOYSA-N | |
| 6776 | |
| 2,6-dihydroxyanthracene-9,10-dione |
Safety and Handling
EINECSNumber : (4)-0704
RTECSNumber : CB6675000
TSCA : Yes