CAS RN 84-60-6
IUPAC Name:
2,6-dihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms:
anthraflavic acid anthraflavin Az-F
Molecular Weight (g/mol):
240.21
Molecular Formula:
C14H8O4
InChi Key:
APAJFZPFBHMFQR-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C(=O)C3=CC(O)=CC=C3C(=O)C2=C1
1
–
1
of
1
results
1
Anthraflavic Acid 97.0+%, TCI America™
CAS: 84-60-6 Molecular Formula: C14H8O4 Molecular Weight (g/mol): 240.214 InChI Key: APAJFZPFBHMFQR-UHFFFAOYSA-N Synonym: 2,6-dihydroxyanthraquinone,anthraflavic acid,anthraflavin,2,6-dihydroxyanthra-9,10-quinone,9,10-anthracenedione, 2,6-dihydroxy,az-f,anthrafravic acid,anthraquinone, 2,6-dihydroxy,2,6-dihydroxy-9,10-anthraquinone,2,6-dihydroxy-anthraquinone PubChem CID: 6776 ChEBI: CHEBI:34250 IUPAC Name: 2,6-dihydroxyanthracene-9,10-dione SMILES: C1=CC2=C(C=C1O)C(=O)C3=C(C2=O)C=C(C=C3)O