missing translation for 'onlineSavingsMsg'
Learn More

Anthraflavic Acid 97.0+%, TCI America™

Catalog No. A18941G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18941G 1 g
A18945G 5 g
A189425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A18941G Supplier TCI America Supplier No. A18941G
Only null left

Chemical Identifiers

CAS 84-60-6
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
InChI Key APAJFZPFBHMFQR-UHFFFAOYSA-N
Synonym 2,6-dihydroxyanthraquinone, anthraflavic acid, anthraflavin, 2,6-dihydroxyanthra-9,10-quinone, 9,10-anthracenedione, 2,6-dihydroxy, az-f, anthrafravic acid, anthraquinone, 2,6-dihydroxy, 2,6-dihydroxy-9,10-anthraquinone, 2,6-dihydroxy-anthraquinone
PubChem CID 6776
ChEBI CHEBI:34250
IUPAC Name 2,6-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C=C1O)C(=O)C3=C(C2=O)C=C(C=C3)O

Specifications

Melting Point 330°C
Quantity 1 g
Formula Weight 240.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Anthraflavic Acid
EINECSNumber (4)-0704
RTECSNumber CB6675000
TSCA Yes