missing translation for 'onlineSavingsMsg'
Learn More

Anthracene Zone Refined (number of passes:30) 99.5+%, TCI America™

Catalog No. A0405ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0405ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0405ZONE Supplier TCI America Supplier No. A04051SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 217°C
Color White
Boiling Point 342°C
Quantity ZONE
UN Number 3077
Formula Weight 178.23
Percent Purity ≥99.5% (GC)
Physical Form Crystals
Chemical Name or Material Anthracene Zone Refined (number of passes:30)
EINECSNumber (4)-0683
RTECSNumber CA9350000
TSCA Yes