missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, Practical, Spectrum™ Chemical

Catalog No. p-7165028
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 kg
100 g
12 kg
2.5 kg
45 kg
500 g
Packaging:
Poly Pail
Catalog No. Quantity Packaging
18608883 100 g Poly Pail
18608887 500 g Poly Pail
18608885 2.5 kg Poly Pail
18608882 10 kg Poly Pail
18608884 12 kg Poly Pail
18608886 45 kg Poly Pail
6 options

Catalog No. 18608883

Supplier: Spectrum Chemical Mfg Cor A1296100GM

Only null left

Chemical Identifiers

CAS 120-12-7

Specifications

CAS Max % 1
Melting Point 215° to 219°C
Boiling Point 342°C
Packaging Poly Pail
Quantity 100 g
Grade Practical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.