missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, Practical, Spectrum™ Chemical

Catalog No. 18608883
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
12 kg
45 kg
Packaging:
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608883 100 g Poly Pail
18608887 500 g Poly Pail
18608885 2.5 kg Poly Pail
18608882 10 kg Poly Pail
18608884 12 kg Poly Pail
18608886 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608883 Supplier Spectrum Chemical Mfg Cor Supplier No. A1296100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 120-12-7

Specifications

CAS Max % 1
Melting Point 215° to 219°C
Boiling Point 342°C
Packaging Poly Pail
Quantity 100 g
Grade Practical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.