missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 99%

Numéro de catalogue. p-7080572
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1020314 25 g
AAA1020322 100 g
AAA1020336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1020314 Fournisseur Thermo Scientific Chemicals Code fournisseur A1020314
Il en reste null

CAS: 120-12-7 | C14H10 | 178.23 g/mol

Anthracene is used in the production of the red dyealizarin and other dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene is used in the production of the red dyealizarin and other dyes.

Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.

Identifiants chimiques

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Spécifications

Melting Point 214°C to 218°C
Density 1.25
Boiling Point 340°C to 342°C
Flash Point 121°C (249°F)
Quantity 25 g
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
Solubility Information Soluble in alcohol,methanol (1.8g/100 g),hexane (0.37g/100 g). Insoluble in water.
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Anthracene
Afficher plus de résultats Afficher moins de résultats
Hazard Category H315-H319-H500
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-371-1
RTECSNumber CA9350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only