missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9-carboxylic acid, 98+%

Catalog No. AAA1404909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404909 10 g
AAA1404918 50 g
AAA1404930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404909 Supplier Thermo Scientific Chemicals Supplier No. A1404909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 723-62-6 | C15H10O2 | 222.24 g/mol

Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.

Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.

Chemical Identifiers

CAS 723-62-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00001257
InChI Key XGWFJBFNAQHLEF-UHFFFAOYSA-N
Synonym 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
PubChem CID 2201
ChEBI CHEBI:34507
IUPAC Name anthracene-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point ∼218°C (decomposition)
Quantity 10 g
Beilstein 1875336
Solubility Information Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene,chloroform,and hydronaphthalenes. Insoluble in water.
Formula Weight 222.24
Percent Purity ≥98%
Chemical Name or Material Anthracene-9-carboxylic acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-964-9
RTECSNumber CB8764000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only