missing translation for 'onlineSavingsMsg'
Learn More

Ansul™ Bulk SPILL-X™ Agents

Catalog No. 17978
Encompass
Change view
Click to view available options
Base:
Citric Acid
Magnesium Oxide
Urea
Color:
Green
Red
Tan
Weight (English):
37 lb.
42 lb.
50 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Base Color Weight (English)
17978 Urea Green 37 lb.
172001A Magnesium Oxide Red 50 lb.
172001C Citric Acid Tan 42 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste material should comply with local, state, and federal regulations.
Catalog No. 17978 Supplier Ansul™ Supplier No. 78443

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste material should comply with local, state, and federal regulations.

For response to larger spills

  • Use a scoop or shovel to apply the agent directly onto the spill
  • Bulk-packed agents can also be used to refill a Spill-Gun™ applicator (not included)
  • Include detailed instructions for responding to a spill

Specifications

Base Urea
Color Green
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Type Spill-X-Fp Agent
For Use With (Application) Small formaldehyde spills such as those in labs and stockrooms
For Use With (Equipment) Spill Gun™ applicator
Includes Detailed instructions for responding to a spill
Weight (English) 37 lb.
Weight (Metric) 16.8 kg