missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ANSI Sign: CONFINED SPACE
Improve workplace safety with non-adhesive mounted signage. The Brady™ Confined Space ANSI Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
Supplier: Brady™ 143680
Description
- This Confined Space Sign has permanently embedded graphics within the sign panel and has excellent outdoor durability
- Brady™ signs feature bright colors, bold text, and intuitive pictographs
- Highly visible and easily understood
- Corner holes provide easy mounting solutions in a variety of environments
Chemical Identifiers
| Not Laminated | |
| Corner Holes | |
| Rectangular |
| 1 Ea. | |
| -40°F | |
| CONFINED SPACE |
Specifications
| Not Laminated | |
| 1 Ea. | |
| 7 in. | |
| -40°F | |
| 10 in. | |
| -40°F to 230°F | |
| CONFINED SPACE |
| Aluminum | |
| Corner Holes | |
| 230°F | |
| 0.035 in. | |
| Rectangular | |
| 10 x 7 in. | |
| Black |