missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-600 Heavy-Duty Cut-Resistant Gloves

Heavy-duty, cut-resistant gloves are ideal for safely handling glass, metal and plastics.

Supplier:  Ansell 8060011090

 View more versions of this product

Catalog No. 19-170-504D


Only null left
Explore available promotions

Alert:

This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Description

Description

  • Safely handles glass, metal and plastics
  • Cut resistant and mechanical performance
  • Aramid fiber knitted liner and natural rubber coating
  • Withstands abrasion, tearing and punctures
  • Latex coating raised crinkle finish provides secure grip in both dry and wet conditions
Specifications

Specifications

ANSI 4
Knit
Blue, Yellow
Specific
Natural Rubber
9
Sheet metal and glass handling, recycling, canning and bottling applications
Aramid Fiber
ActivArmr™ 80-600
25 cm
EN 407, CE 0493, EN 420:2003 + A1:2009; Category III
Latex
ANSI A2
No
Cut-resistant Gloves
EN 388:3444, EN 388:3444C
Knit
80-600-9
9.6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions