missing translation for 'onlineSavingsMsg'
Learn More

ANS-NH4 (=Ammonium 8-Anilino-1-naphthalenesulfonate) 95.0+%, TCI America™

Catalog No. p-7118273
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53511G 1 g
A535110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53511G Supplier TCI America Supplier No. A53511G
Only null left

Chemical Identifiers

CAS 28836-03-5
Molecular Formula C16H16N2O3S
Molecular Weight (g/mol) 316.38
MDL Number MFCD00012560
InChI Key IPBNQYLKHUNLQE-UHFFFAOYSA-N
Synonym ammonium 8-phenylamino naphthalene-1-sulfonate, 8-anilino-1-naphthalenesulfonic acid ammonium salt, unii-gt7mn8dkd7, gt7mn8dkd7, ammonium 8-anilino-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 8-phenylamino-, monoammonium salt, 1-naphthalenesulfonic acid, 8-phenylamino-, ammonium salt 1:1, ansa, 1-anilinonaphthalene-8-sulfonic acid ammonium salt, 82-76-8 parent
PubChem CID 120066
IUPAC Name ammonium 8-(phenylamino)naphthalene-1-sulfonate
SMILES [NH4+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1

Specifications

Melting Point 237°C
Quantity 1 g
Formula Weight 316.38
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material ANS-NH4 (=Ammonium 8-Anilino-1-naphthalenesulfonate) [Hydrophobic fluorescent probe]