missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572527
Description
ANP Polyclonal Antibody for Flow, IHC (P), Western Blot
The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.Specifications
| ANP | |
| Polyclonal | |
| Unconjugated | |
| NPPA | |
| Anf; ANP; ATFB6; atrial natriuretic factor; Atrial natriuretic peptide; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; ATRST2; Auriculin-A; Auriculin-B; Cardiodilatin; Cardiodilatin-related peptide; cardionatrin; CDD; CDD-ANF; CDP; natriuretic peptide A; Natriuretic peptide precursor A (pronatriodilatin, also Anf, Pnd); natriuretic peptide precursor A variant 1; natriuretic peptide precursor A variant NPPA-M1; natriuretic peptide precursor A variant NPPA-M2; natriuretic peptide precursor A variant NPPA-M3; natriuretic peptide precursor type A; natriuretic peptide type A; natriuretic peptides A; Nppa; Pnd; Prepronatriodilatin; Pronatriodilatin; RATANF; urodilatin | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 230899, 4878 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.46 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P01160, P05125 | |
| NPPA | |
| KLH conjugated synthetic peptide between 30-56 amino acids from the N-terminal region of human NPPA. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |