missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Anode Buffer Reservoir Jar

Catalog No. 5402
Encompass
Change view
Click to view available options
Quantity:
1 jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5402 1 jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5402 Supplier Applied Biosystems™ Supplier No. LS5402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Buffer Reservoir is for use with the 3100-Avant™ Genetic Analyzer.

This Buffer Reservoir is for use with the 3100-Avant™ Genetic Analyzer. It attaches to the lower polymer block, holding the anodic buffer.

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
No. of Reactions 13
Product Type Anode Buffer Reservoir Jar
Quantity 1 jar
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.