missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Anode Buffer Container (ABC) for 3500 Series Systems for Protein Quality Analysis

Catalog No. A31278
Encompass
Change view
Click to view available options
Quantity:
4 Containers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31278 4 Containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31278 Supplier Applied Biosystems™ Supplier No. A31278

missing translation for 'validMainframeMsg'

Includes

Contains 4 anode buffer containers.

The anode buffer container (ABC) contains 1X running buffer to support protein quality analysis applications on the Applied Biosystems™ 3500 series genetic analyzers for protein quality analysis.

  • Each package includes four containers

Specifications

Content And Storage Contains 4 Anode Buffer containers.
Store at 2–8°C until ready to use. Refer to the expiration date on the label.
For Use With (Equipment) 3500 Genetic Analyzer for Protein Quality Analysis, 3500xL Genetic Analyzer for Protein Quality Analysis
For Use With (Application) Sequencing
Product Type Anode Buffer Container
Quantity 4 Containers
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.