missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANO5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572534
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572534 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572534 Supplier Invitrogen™ Supplier No. PA572534
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

ANO5 Polyclonal Antibody for Western Blot, IHC (P)

Ano5 does not exhibit calcium-activated chloride channel (CaCC) activity. The Ano5 gene encodes a member of the anoctamin family of transmembrane proteins. The Ano5 protein is likely a calcium activated chloride channel. Mutations in the Ano5 gene have been associated with gnathodiaphyseal dysplasia. Alternatively spliced transcript variants have been described for Ano5. Diseases associated with ANO5 include Gnathodiaphyseal Dysplasia and Miyoshi Muscular Dystrophy 3.

Specifications

Antigen ANO5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ANO5
Gene Accession No. Q75UR0, Q75V66
Gene Alias 9330162L24; Ano5; anoctamin 5; anoctamin-5; GDD1; gnathodiaphyseal dysplasia 1 protein; Gnathodiaphyseal dysplasia 1 protein homolog; integral membrane protein GDD1; LGMD2L; RGD1564265; Tmem16e; transmembrane protein 16E
Gene Symbols ANO5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 792-819 amino acids from the C-terminal region of human ANO5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203859, 233246
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less