missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin V Recombinant Rabbit Monoclonal Antibody (23GB650)
GREENER_CHOICE

Catalog No. MA554184
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554184 100 μL
MA554185 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554184 Supplier Invitrogen™ Supplier No. MA554184
Only null left

Rabbit Recombinant Monoclonal Antibody

Annexin V Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.

Specifications

Antigen Annexin V
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB650
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Anxa5
Gene Accession No. P08758, P14668, P48036
Gene Alias Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Gene Symbols Anxa5
Host Species Rabbit
Immunogen A synthetic peptide of human Annexin V (301-320AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11747, 25673, 308
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less