missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A7 Recombinant Rabbit Monoclonal Antibody (23GB1235)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | Annexin A7 |
|---|---|
| Clone | 23GB1235 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
Description
Annexin A7 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima.Specifications
| Annexin A7 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ANXA7 | |
| AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN | |
| ANXA7 | |
| Primary | |
| Affinity chromatography |
| 23GB1235 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20073, Q07076 | |
| 11750, 155423, 310 | |
| A synthesized peptide derived from human Annexin A7 (1-150AA). | |
| Antibody |