missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A3 Monoclonal Antibody (2#)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA565976
Description
Annexin A3 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.Chemical Identifiers
| Annexin A3 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O35639, P12429, P14669 | |
| 11745, 25291, 306 | |
| Recombinant Annexin A3 (ANXA3) (Ala26-Ala160). | |
| Antibody |
| 2# | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human, Mouse, Rat | |
| ANXA3 | |
| 35-alpha calcimedin; Annexin 3; annexin A3; Annexin III; Annexin III (Lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin3; annexin-3; ANX3; ANXA 3; ANXA3; calcimedin 35-alpha; inositol 1,2-cyclic phosphate 2-phosphohydrolase; LC3; Lipocortin III; LRRGT00047; PAP-III; Placental anticoagulant protein III | |
| ANXA3 | |
| Primary | |
| Protein A/G |
Specifications
| Annexin A3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O35639, P12429, P14669 | |
| ANXA3 | |
| Recombinant Annexin A3 (ANXA3) (Ala26-Ala160). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 2# | |
| Unconjugated | |
| ANXA3 | |
| 35-alpha calcimedin; Annexin 3; annexin A3; Annexin III; Annexin III (Lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin3; annexin-3; ANX3; ANXA 3; ANXA3; calcimedin 35-alpha; inositol 1,2-cyclic phosphate 2-phosphohydrolase; LC3; Lipocortin III; LRRGT00047; PAP-III; Placental anticoagulant protein III | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11745, 25291, 306 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |