missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557806
Description
Immunogen sequence: ETKKVVLVDG KTKDHRRSSS SRSQDSAEGQ DGQVPEQFSG LLHGSSPVCE VGQDPFQLLC TAGQSHPDGS PQQGACHKAS MQLEETGVHA PG Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 72%.
Regulator of different signaling pathways. Regulates EPHA8 receptor tyrosine kinase signaling to control cell migration and neurite retraction.Specifications
| ANKS1 | |
| Polyclonal | |
| Unconjugated | |
| ANKS1A | |
| ANKS1; ANKS1A; ankyrin repeat and SAM domain containing 1; ankyrin repeat and SAM domain-containing protein 1A; ankyrin repeat and sterile alpha motif domain containing 1; ankyrin repeat and sterile alpha motif domain containing 1A; KIAA0229; ODIN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92625 | |
| ANKS1A | |
| Recombinant protein corresponding to Human ANKS1. Recombinant protein control fragment (Product #RP-94614). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |