missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisonitrile 98.0+%, TCI America™
$85.75 - $337.40
Chemical Identifiers
| CAS | 874-90-8 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00001818 |
| InChI Key | XDJAAZYHCCRJOK-UHFFFAOYSA-N |
| Synonym | anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile |
| PubChem CID | 70129 |
| IUPAC Name | 4-methoxybenzonitrile |
| SMILES | COC1=CC=C(C=C1)C#N |
Chemical Identifiers
| 874-90-8 | |
| 133.15 | |
| XDJAAZYHCCRJOK-UHFFFAOYSA-N | |
| 70129 | |
| COC1=CC=C(C=C1)C#N |
| C8H7NO | |
| MFCD00001818 | |
| anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile | |
| 4-methoxybenzonitrile |
Specifications
| 874-90-8 | |
| White-Yellow | |
| C8H7NO | |
| 5 g | |
| anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile | |
| COC1=CC=C(C=C1)C#N | |
| 133.15 | |
| 133.15 | |
| Crystalline Powder |
| 60°C | |
| 137°C | |
| MFCD00001818 | |
| 3439 | |
| XDJAAZYHCCRJOK-UHFFFAOYSA-N | |
| 4-methoxybenzonitrile | |
| 70129 | |
| ≥98.0% (GC) | |
| Anisonitrile |
Safety and Handling
TSCA : Yes