missing translation for 'onlineSavingsMsg'
Learn More

Anisonitrile, 99%

Catalog No. AC158050250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158050250 25 g Glass bottle
AC158051000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158050250 Supplier Thermo Scientific Chemicals Supplier No. 158050250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 874-90-8 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-90-8
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001818
InChI Key XDJAAZYHCCRJOK-UHFFFAOYSA-N
Synonym anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
PubChem CID 70129
IUPAC Name 4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1)C#N

Specifications

Melting Point 57°C to 60°C
Color White to Beige
Boiling Point 256°C to 257°C (765.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CN
Quantity 25 g
Beilstein 10, 168
Solubility Information Solubility in water: soluble
Formula Weight 133.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Anisonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-871-6
TSCA TSCA

RUO – Research Use Only