missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisomycin 97.0+%, TCI America™
$355.05 - $1042.34
Chemical Identifiers
| CAS | 22862-76-6 |
|---|---|
| Molecular Formula | C14H19NO4 |
| Molecular Weight (g/mol) | 265.31 |
| MDL Number | MFCD00077650 |
| InChI Key | YKJYKKNCCRKFSL-BFHYXJOUSA-N |
| Synonym | Flagecidin, (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol 3-Acetate |
| PubChem CID | 253602 |
| ChEBI | CHEBI:338412 |
| IUPAC Name | (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate |
| SMILES | COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A261325MG
|
TCI America
A261325MG |
25 mg |
Each for $355.05
|
|
|||||
|
A2613100MG
|
TCI America
A2613100MG |
100 mg |
Each for $1,042.34
|
|
|||||
Chemical Identifiers
| 22862-76-6 | |
| 265.31 | |
| YKJYKKNCCRKFSL-BFHYXJOUSA-N | |
| 253602 | |
| (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate |
| C14H19NO4 | |
| MFCD00077650 | |
| Flagecidin, (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol 3-Acetate | |
| CHEBI:338412 | |
| COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1 |
Specifications
| Anisomycin | |
| 22862-76-6 | |
| White | |
| C14H19NO4 | |
| 3462 | |
| YKJYKKNCCRKFSL-BFHYXJOUSA-N | |
| (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate | |
| 253602 | |
| 265.31 | |
| Anisomycin |
| Crystalline Powder | |
| 144°C | |
| 25 mg | |
| MFCD00077650 | |
| Flagecidin, (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol 3-Acetate | |
| COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1 | |
| 265.31 | |
| CHEBI:338412 | |
| ≥97.0% (GC) |
Safety and Handling
RTECSNumber : BZ9800000
TSCA : No
Recommended Storage : Freezer (-20 °C)