missing translation for 'onlineSavingsMsg'
Learn More

Anisomycin, 97%

CAS: 22862-76-6 | C14H19NO4 | 265.31 g/mol

Supplier:  Thermo Scientific Chemicals J62964MF

Catalog No. AAJ62964MF


Only null left

Description

Description

A protein synthesis inhibitor. Causes apoptosis of PC12 Cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A protein synthesis inhibitor
  • Causes apoptosis of PC12 Cells
Specifications

Specifications

Anisomycin
Powder
140°C to 141°C
50 mg
MFCD00077650
20705
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)-3-pyrrolidinyl acetate; Flagecidin
YKJYKKNCCRKFSL-BFHYXJOUSA-N
(2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
253602
265.31
Anisomycin
22862-76-6
White
C14H19NO4
UN3462
14,670
Soluble in water at 2mg/ml,in DMSO at 20mg/ml,and in methanol at 20mg/ml
COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1
265.31
CHEBI:338412
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

EINECSNumber : 245-269-7

RTECSNumber : BZ9800000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only