missing translation for 'onlineSavingsMsg'
Learn More

Anisole 99.0+%, TCI America™

Catalog No. A049225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A049225G 25 g
A0492500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A049225G Supplier TCI America Supplier No. A049225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-66-3
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00008354
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1

Specifications

Melting Point -37°C
Color Colorless
Boiling Point 154°C
Quantity 25 g
UN Number 2222
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Anisole
EINECSNumber (3)-0556
RTECSNumber BZ8050000
TSCA Yes