missing translation for 'onlineSavingsMsg'
Learn More

Anisole, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187484
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019907 5 mL
111019073 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019907 Supplier Merck Emd Millipore Supplier No. 961095MLF
Only null left

Anisole is a colorless liquid , which can be obtained upon heating anisic acid with an excess of barium hydroxide.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 100-66-3
Ignition Point 887°F
Vapor Pressure 10 mmHg (42.2°C)
Vapor Density 3.7 (vs. air)
Density 0.995 g/mL (at 25°C)
Flash Point 43°C
Physical Form Neat
Linear Formula CH3 OC6H5
MDL Number MFCD00008354
Melting Point -37°C
Boiling Point 154°C
Synonym Methoxybenzene; Methyl phenyl ether
Type Analytical Standard
Quantity 5 mL
Formula Weight 108.14
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less