Learn More
Anisil, 98+%
CAS: 1226-42-2 | C16H14O4 | 270.284 g/mol
Supplier: Thermo Scientific Chemicals A1059414
Description
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Anisil | |
| 132°C to 134°C | |
| MFCD00008405 | |
| 531345 | |
| Insoluble in water. | |
| COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC | |
| 270.284 | |
| 270.29 |
| 1226-42-2 | |
| C16H14O4 | |
| 25 g | |
| 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
| YNANGXWUZWWFKX-UHFFFAOYSA-N | |
| 1,2-bis(4-methoxyphenyl)ethane-1,2-dione | |
| 71043 | |
| ≥98% |
Chemical Identifiers
| 1226-42-2 | |
| 270.284 | |
| YNANGXWUZWWFKX-UHFFFAOYSA-N | |
| 71043 | |
| COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC |
| C16H14O4 | |
| MFCD00008405 | |
| 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
| 1,2-bis(4-methoxyphenyl)ethane-1,2-dione |
Safety and Handling
EINECSNumber : 214-960-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only