missing translation for 'onlineSavingsMsg'
Learn More

Anions, MilliporeSigma™ Supelco™

Catalog No. 111002610
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002610 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002610 Supplier Merck Emd Millipore Supplier No. QC136420ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental and Food and Beverages

Specifications

CAS 7778-80-5
Type Certified Reference Material
Quantity 20 mL
Grade Certified Reference Material