Learn More
Stoelting™ Animal Markers
Animal Markers allow for easy, long-lasting identification of laboratory animals.
$227.59 - $630.30
| Catalog Number | Mfr. No. | Size | Model | Includes | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Model | Includes | Price | Quantity & Availability | |||
|
10000646
|
Stoelting™
50451 |
Fine tip | Black | Pack of 3 |
Pack of 3 Each for $630.30
|
|
|||
|
10000645
|
Stoelting™
50450 |
Standard tip | Black | Pack of 2 |
Pack of 2 Each for $525.24
|
|
|||
|
10001083
|
Stoelting™
STG504501 |
Standard tip | Black | Single Packaging |
Each for $280.13
|
|
|||
|
10001073
|
Stoelting™
STG504401 |
Standard tip | Blue | Single Packaging |
Each for $280.13
|
|
|||
|
10001074
|
Stoelting™
504411 |
Standard tip | Green | Single Packaging |
Each for $280.13
|
|
|||
|
10001082
|
Stoelting™
504491 |
Fine tip | Orange | Single Packaging |
Each for $227.59
|
|
|||
|
10001077
|
Stoelting™
504441 |
Standard tip | Orange | Single Packaging |
Each for $280.13
|
|
|||
|
10000642
|
Stoelting™
50447 |
Fine tip | Red | Pack of 3 |
Pack of 3 Each for $630.30
|
|
|||
|
10001075
|
Stoelting™
504421 |
Standard tip | Red | Single Packaging |
Each for $280.13
|
|
|||
|
10001076
|
Stoelting™
504431 |
Standard tip | Yellow | Single Packaging |
Each for $280.13
|
|
|||
Description
- Nontoxic and devoid of any hazardous materials
- A single marker can be used to mark 300 to 500 rats (depending upon animal size and/or applied area)
- Once applied, color remains on fur for 6 to 12 weeks; does not work on bare skin
- Broad and fine tip markers available
Specifications
| Neuroscience | |
| Rodent |