Learn More
Stoelting™ Animal Markers
Catalog No. 10000646
Change view
Click to view available options
Size:
Fine tip
Standard tip
Model:
Black
Blue
Green
Orange
Red
Yellow
Includes:
Pack of 2
Pack of 3
Single Packaging
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Model Includes
10000646 Fine tip Black Pack of 3
10000645 Standard tip Black Pack of 2
10001083 Standard tip Black Single Packaging
10001073 Standard tip Blue Single Packaging
10001074 Standard tip Green Single Packaging
10001082 Fine tip Orange Single Packaging
10001077 Standard tip Orange Single Packaging
10000642 Fine tip Red Pack of 3
10001075 Standard tip Red Single Packaging
10001076 Standard tip Yellow Single Packaging
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 10000646 Supplier Stoelting™ Supplier No. 50451
Animal Markers allow for easy, long-lasting identification of laboratory animals.
  • Nontoxic and devoid of any hazardous materials
  • A single marker can be used to mark 300 to 500 rats (depending upon animal size and/or applied area)
  • Once applied, color remains on fur for 6 to 12 weeks; does not work on bare skin
  • Broad and fine tip markers available
Specifications
For Use With (Application) Neuroscience
Size Fine tip
Animal Rodent
Model Black
Includes Pack of 3