missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anillin Monoclonal Antibody (KAA135_9A4), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Specifications
| Antigen | Anillin |
|---|---|
| Clone | KAA135_9A4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: TPNTKAIQER
Anillin Monoclonal antibody specifically detects Anillin in Human samples. It is validated for Western BlotSpecifications
| Anillin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1110037A17Rik; 2900037I21Rik; actin-binding protein anillin; Anillin; anillin actin binding protein; anillin, actin binding protein; anillin, actin binding protein (scraps homolog, Drosophila); Anln; C78101; DKFZp779A055; FSGS8; RGD1561367; scra; Scraps | |
| Human ANLN peptide (immunogen range is 401-410). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ANLN |
| KAA135_9A4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NQW6 | |
| 54443 | |
| IgG1 | |
| Protein A | |
| Antibody |