missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydroiodide 98.0+%, TCI America™

Catalog No. A27781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27781G 1 g
A27785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27781G Supplier TCI America Supplier No. A27781G

Chemical Identifiers

CAS 45497-73-2
Molecular Formula C6H8IN
Molecular Weight (g/mol) 221.04
MDL Number MFCD21607646
InChI Key KFQARYBEAKAXIC-UHFFFAOYSA-N
Synonym Anilinium Iodide
PubChem CID 24192279
IUPAC Name aniline hydroiodide
SMILES I.NC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 221.04
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Aniline Hydroiodide