accessibility menu, dialog, popup

UserName

NH2HI

CAS RN 45497-73-2

IUPAC Name: aniline hydroiodide
Synonyms: aniline hydroiodide
Molecular Weight (g/mol): 221.04
Molecular Formula: C6H8IN
InChi Key: KFQARYBEAKAXIC-UHFFFAOYSA-N
SMILES: I.NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)