missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydrogen Phthalate 98.0+%, TCI America™

Catalog No. P028425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P028425G Supplier TCI America Supplier No. P028425G

Chemical Identifiers

CAS 50930-79-5
Molecular Formula C14H13NO4
Molecular Weight (g/mol) 259.261
MDL Number MFCD00042034
InChI Key QGPZXNSBZMHHSR-UHFFFAOYSA-N
Synonym aniline hydrogen phthalate, aniline phthalate, aniline; phthalic acid, 1,2-benzenedicarboxylic acid, compd. with benzenamine 1:1, c8h6o4.c6h7n, phthalic acid, compound with aniline 1:1, aniline hydrogenphthalate, aniline hydrogen phtalate, aniline phthalate solution, phthalic acid aniline salt
PubChem CID 170940
IUPAC Name aniline;phthalic acid
SMILES C1=CC=C(C=C1)N.C1=CC=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 259.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aniline Hydrogen Phthalate